Skip to Content
Merck
All Photos(1)

Documents

1.08122

Supelco

Bromophenol blue

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Bromophenol blue, 3,3′,5,5′-Tetrabromophenolsulfonphthalein, BPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
MDL number:
UNSPSC Code:
12171502
EC Index Number:
204-086-2
NACRES:
NA.21

product name

Bromophenol blue, indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

loss

≤1% loss on drying, 110°C

visual transition interval

3.0-4.6, greenish yellow to blue-violet

mp

270-273 °C (decomposition)

bulk density

730 kg/m3

storage temp.

2-30°C

InChI

1S/C19H10Br4O5S/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

UDSAIICHUKSCKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Non-targeted analysis and toxicity prediction for evaluation of photocatalytic membrane distillation removing organic contaminants from hypersaline oil and gas field-produced water.: The study uses 2-Butanone in experimental setups to assess the removal of hazardous organic compounds in water treatment processes, highlighting its role in environmental safety and sustainability (Delanka-Pedige et al., 2024).

  • Lipid and volatile profiles of Finnish oat batches of pure cultivars: Effect of storage on the volatile formation.: This research utilizes 2-Butanone for the extraction and analysis of volatile compounds in oats, which helps in understanding the effects of storage conditions on food quality and safety (Puganen et al., 2024).



Analysis Note

Transition range: pH 3.0 - pH 4.6greenish yellow - blue violet
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 1
Transition range (according to ACS): passes test
Sensitivity test: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service