Skip to Content
Merck
All Photos(1)

Key Documents

1.07041

Supelco

Diethylamine

for analysis EMPARTA® ACS

Synonym(s):

Diethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352116
grade:
ACS reagent
Pricing and availability is not currently available.

grade

ACS reagent

storage temp.

20-25°C

SMILES string

N(CC)CC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service