Skip to Content
Merck
All Photos(1)

Key Documents

1.05700

Millipore

L-Lysine monohydrochloride

for biochemistry

Synonym(s):

L-Lysine monohydrochloride, Lys

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€45.30
1 KG
€204.00
10 KG
€1,670.00

€45.30


Please contact Customer Service for Availability


Select a Size

Change View
100 G
€45.30
1 KG
€204.00
10 KG
€1,670.00

About This Item

Empirical Formula (Hill Notation):
C6H15ClN2O2
CAS Number:
Molecular Weight:
182.65
MDL number:
UNSPSC Code:
12352209
EC Index Number:
211-519-9
NACRES:
NA.21

€45.30


Please contact Customer Service for Availability

vapor pressure

<1 Pa ( 20 °C)

Quality Level

potency

10000 mg/kg LD50, oral (Rat)

pH

5.5-6.0 (20 °C, 100 g/L in H2O)

mp

262 °C

density

1.28 g/cm3 at 20 °C

bulk density

360 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3][C@@H](CCCCN)C(=O)O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine powder
Appearance of solution (1 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +20.3 to +21.3
UV-Absorption(260 nm; 1 mol/l; 1 cm; water): ≤ 0.20
UV-Absorption(280 nm; 1 mol/l; 1 cm; water): ≤ 0.25
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.4 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service