Skip to Content
Merck
All Photos(1)

Key Documents

1.05467

Millipore

Selective agar for pathogenic fungi

for the isolation of pathogenic fungi (contains Cycloheximide)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

powder (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

6.8-7.0 (25 °C, 33 g/L in H2O)

solubility

33 g/L

bulk density

670 kg/m3

application(s)

clinical
microbiology

suitability

molds

General description

for the isolation of pathogenic fungi (contains Cycloheximide)

Analysis Note

Appearance (clearness)clearAppearance (colour)yellowish-brownpH-value (25 °C)6.7 - 7.1Growth (Trichophyton mentagrophytes ATCC 18748)good to very goodGrowth (Trichophyton rubrum ATCC 28188)fair to goodGrowth (Microsporum gallinae ATCC 12108)fair to goodGrowth (Trichophyton ajelloi ATCC 28454)fair to goodGrowth (Microsporum canis ATCC 36299)good to very goodGrowth (Geotrichum candidum DSM 1240)good to very goodGrowth (Candida albicans ATCC 10231 (WDCM 00054))good to very goodGrowth (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053))none to poorGrowth (Penicillium commune ATCC 10428)none to poorGrowth (Bacillus cereus ATCC 11778 (WDCM 00001))noneIncubation: 7 days; 28 °C, aerobic.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service