Skip to Content
Merck
All Photos(1)

Documents

104551

Sigma-Aldrich

ACC2 Inhibitor, CD-017-0191

The ACC2 Inhibitor, CD-017-0191 controls the biological activity of ACC2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

ACC2 Inhibitor, CD-017-0191, 2-((2-(4-isopropoxyphenyl)-5-methyloxazol-4-yl)methylthio)oxazolo[4,5-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N3O3S
Molecular Weight:
381.45
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable and selective inhibitor of human Acetyl-CoA carboxylase 2 (ACC2) (IC50 = 2.8 µM), which does not inhibit human ACC1 at all at concentrations up to 100 µM, in vitro. It is also shown to inhibit the growth of yeast (IC50 = 3.8 µM), in which the native ACC1 gene is replaced with the human ACC2 strain; but not with the human ACC1 strain at concentrations ≤ 100 µM. This compound is not demonstrated to be toxic to human cells in culture at concentrations up to 10 µM.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Marjanovic, J., et al. 2010 Proc. Natl. Acad. Sci.107, 9093.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service