Skip to Content
Merck
All Photos(1)

Documents

1.04208

Sigma-Aldrich

Brilliant green (hydrogen sulfate) (C.I. 42040)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid

Quality Level

potency

300-400 mg/kg LD50, oral (Rat)

composition

Dye, ≥95% spectrophotometric assay

pH

2.5 (25 °C, 10 g/L in H2O)

mp

210 °C (decomposition)

solubility

100 g/L

bulk density

500‑600 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant green (hydrogen sulfate) (C.I. 42040) for microscopy Certistain®, is a dry staining dye that is used to prepare a staining solution makes target structures in bacteriological specimen materials evaluable for diagnostic purposes. In the area of bacteriology, among others the Gram staining method, the Ziehl-Neelsen staining method, and the spore-staining method according to Rakette are used to yield a differentiated image of pathogens. This can be done either by using eosin Y as a counterstain or by modification according to Wirtz using safranine O solution. By this method, spores are always stained green and the cell elements always red. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 95 %
Identity
- Hydrogen sulfate (as sulfur):
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50 %): 628 - 632 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 2210 - 2350
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service