Skip to Content
Merck
All Photos(1)

Key Documents

1.04036

Millipore

E.coli 0157:H7-Agar

for microbiology Fluorocult®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

sterility

sterile

form

powder (dehydrated (DCM))

manufacturer/tradename

FluoroCult®

technique(s)

microbiological culture: suitable

pH

7.2-7.6 (25 °C, 55 g/L in H2O, after autoclaving)

solubility

55 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

enterovirulent E. coli (EEC)

Analysis Note

Appearance (clearness): clear
Appearance (colour): green to blue-green
pH-value (25 °C): 7.2 - 7.6
Growth (Escherichia coli ATCC 35150 (O157)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): fair to very good
Growth (Enterobacter aerogenes ATCC 13048): good to very good
Growth (Proteus mirabilis ATCC 14273): good to very good
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)): good to very good
Growth (Shigella sonnei ATCC 11060): good to very good
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): none
Colony colour (Escherichia coli ATCC 35150 (O157)): colourless
Colony colour (Escherichia coli ATCC 25922 (WDCM 00013)): yellow
Colony colour (Enterobacter aerogenes ATCC 13048): yellow
Colony colour (Proteus mirabilis ATCC 14273): brown
Colony colour (Shigella sonnei ATCC 11060): colourless
Colonies (Salmonella typhimurium ATCC 14028 (WDCM 00031)): yellow with black centre
Fluorescence (Escherichia coli ATCC 35150 (O157)): -
Fluorescence (Escherichia coli ATCC 25922 (WDCM 00013)): +
Fluorescence (Enterobacter aerogenes ATCC 13048): -
Fluorescence (Shigella sonnei ATCC 11060): +
Incubation: 24 hrs.; 35 °C; aerobic.

Legal Information

FLUOROCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service