Skip to Content
Merck
All Photos(1)

Key Documents

1035-HK

Sigma-Aldrich

GP anti GLP-1 (Active) Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1035-HK
€347.00

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1035-HK
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service