Skip to Content
Merck
All Photos(1)

Documents

07-318

Sigma-Aldrich

Anti-TBX2 Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... TBX2(6909)

Specificity

TBX2

Immunogen

GST fusion protein corresponding to residues 554-702 of human TBX2

Application

Detect TBX2 also known as T-box 2 with Anti-TBX2 Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in IP & WB.

Quality

routinely evaluated by immunoblot on RIPA lysates from MCF-7 human breast adenocarcinoma cells

Target description

74kDa

Physical form

antiserum with 0.05% sodium azide before the addition of glycerol to 30%

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Aparicio-Blanco et al.
Drug delivery and translational research (2024-05-13)
Brain drug delivery is severely hindered by the presence of the blood-brain barrier (BBB). Its functionality relies on the interactions of the brain endothelial cells with additional cellular constituents, including pericytes, astrocytes, neurons, or microglia. To boost brain drug delivery
J J Jacobs et al.
Nature genetics, 26(3), 291-299 (2000-11-04)
To identify new immortalizing genes with potential roles in tumorigenesis, we performed a genetic screen aimed to bypass the rapid and tight senescence arrest of primary fibroblasts deficient for the oncogene Bmi1. We identified the T-box member TBX2 as a
Timo H Lüdtke et al.
Respiratory research, 22(1), 85-85 (2021-03-19)
Tbx2 encodes a transcriptional repressor implicated in the development of numerous organs in mouse. During lung development TBX2 maintains the proliferation of mesenchymal progenitors, and hence, epithelial proliferation and branching morphogenesis. The pro-proliferative function was traced to direct repression of
K L Redmond et al.
Oncogene, 29(22), 3252-3262 (2010-03-30)
T-box 2 (TBX2) is a transcription factor involved in mammary development and is known to be overexpressed in a subset of aggressive breast cancers. TBX2 has previously been shown to repress growth control genes such as p14(ARF) and p21(WAF1/cip1). In
Nurullah Aydoğdu et al.
Development (Cambridge, England), 145(23) (2018-11-28)
The organized array of smooth muscle cells (SMCs) and fibroblasts in the walls of visceral tubular organs arises by patterning and differentiation of mesenchymal progenitors surrounding the epithelial lumen. Here, we show that the TBX2 and TBX3 transcription factors have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service