Skip to Content
Merck
All Photos(1)

Key Documents

05-986

Sigma-Aldrich

Anti-JNK2 Antibody

Upstate®, from mouse

Synonym(s):

Anti-JNK-55, Anti-JNK2, Anti-JNK2A, Anti-JNK2ALPHA, Anti-JNK2B, Anti-JNK2BETA, Anti-PRKM9, Anti-SAPK, Anti-SAPK1a, Anti-p54a, Anti-p54aSAPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

monoclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MAPK9(5601)

Specificity

Human, Mouse and Rat

Immunogen

Recombinant human JNK2 protein

Application

Detect JNK2 using this Anti-JNK2 Antibody validated for use in ELISA & WB.
Research Category
Signaling
Research Sub Category
MAP Kinases

Quality

Routinely evaluated by immunoblot.

Target description

46 and/or 54 kDa

Physical form

ImmunoAffinity Purified

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin-Ichiro Hagiwara et al.
PloS one, 13(9), e0203704-e0203704 (2018-09-08)
Functional gastrointestinal disorders (FGIDs) are characterized by dysregulated gut-brain interactions. Emerging evidence shows that low-grade mucosal inflammation and immune activation contribute to FGIDs, including functional dyspepsia (FD). Stress plays an important role in the onset of FD symptoms. In human
S Gupta et al.
Science (New York, N.Y.), 267(5196), 389-393 (1995-01-20)
Treatment of cells with pro-inflammatory cytokines or ultraviolet radiation causes activation of the c-Jun NH2-terminal protein kinase (JNK). Activating transcription factor-2 (ATF2) was found to be a target of the JNK signal transduction pathway. ATF2 was phosphorylated by JNK on
S Gupta et al.
The EMBO journal, 15(11), 2760-2770 (1996-06-03)
The JNK protein kinase is a member of the MAP kinase group that is activated in response to dual phosphorylation on threonine and tyrosine. Ten JNK isoforms were identified in human brain by molecular cloning. These protein kinases correspond to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service