Skip to Content
Merck
All Photos(4)

Key Documents

05-745R-S

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, Trial Size

clone 15-10C-E4, Upstate®, from rabbit

Synonym(s):

H3K4me3, Histone H3 (tri methyl K4)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μL
€112.00

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μL
€112.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15-10C-E4, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
ELISA: suitable
dot blot: suitable
inhibition assay: suitable (peptide)
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys4)

Gene Information

human ... H3F3B(3021)

General description

Recombinant version of Cat. # 05-745, clone MC315

Specificity

Broad species cross-reactivity expected due to sequence homology
Recognizes Histone H3 containing trimethyl-lysine 4 and, to a lesser extent, dimethyl-lysine 4. See certificate of analysis for further details.

Immunogen

BSA-conjugated, synthetic peptide containing the sequence …RTme3KQT… in which me3K corresponds to trimethyl-lysine 4 of human histone H3.

Application

ChIP-seq Analysis:
Representative lot data. Chromatin immunoprecipitation was performed using the Magna ChIP HiSens kit (cat# 17-10460), 3 µL of Anti-trimethyl-Histone H3 (Lys4) antibody (cat# 05-745R-S), 20 µL Protein A/G beads, and 1e6 crosslinked HeLa cell chromatin followed by DNA purification using magnetic beads. Libraries were prepared from Input and ChIP DNA samples using standard protocols with Illumina barcoded adapters, and analyzed on Illumina HiSeq instrument. An excess of eighteen million reads from FastQ files were mapped using Bowtie (http://bowtie-bio.sourceforge.net/manual.shtml) following TagDust (http://genome.gsc.riken.jp/osc/english/dataresource/) tag removal. Peaks were identified using MACS (http://luelab.dfci.harvard.edu/MACS/), with peaks and reads visualized as a custom track in UCSC Genome Browser (http://genome.ucsc.edu) from BigWig and BED files. The highest 25% of peaks identified in the 05-745R and 07-473 datasets showed 99% overlap with peaks identified in the ENCODE H3K4me3 BROAD Histone track for HeLa S3.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, Recombinant, Trial Size (rabbit monoclonal antibody) proven in WB, ChIP, ChIP-seq, ELISA, PIA, Mplex, DB to detect trimethyl-Histone H3 (Lys4) also known as H3K4me3.

Quality

Routinely evaluated by immunoblot on HeLa nuclear extract

Target description

17 kDa

Linkage

Replaces: 05-745

Physical form

Format: Purified
Protein A purified, recombinant antibody in 60 mM Tris, 90 mM NaCl, 0.03% NaN3, 60 mM Glycine, pH 7.5, 40% Glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Bee Choi et al.
Science advances, 7(44), eabk2775-eabk2775 (2021-10-30)
[Figure: see text].

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service