Skip to Content
Merck
All Photos(4)

Key Documents

05-665-AF488

Sigma-Aldrich

Anti-Active-β-Catenin Antibody, clone 8E7, Alexa Fluor 488

clone 8E7, from mouse, ALEXA FLUOR 488

Synonym(s):

Catenin beta-1, Beta-catenin, Active-β-Catenin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€547.00

€547.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€547.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€547.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

8E7, monoclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

zebrafish (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)
mouse ... Ctnnb1(12387)
rat ... Ctnnb1(84353)
zebrafish ... Ctnnb1(30265)

Related Categories

General description

Catenin beta-1 (UniProt P35222; also known as beta-Catenin) is encoded by the CTNNB1 (also known as CTNNB, MRD19) gene (ORF OK/SW-cl.35, PRO2286; Gene ID 1499) in human. beta-Catenin is a multifunctional protein involved in the regulation of cell–cell adhesion and in the control of the Wnt signaling pathway. beta-Catenin contains in its central region 12 armadillo repeats that mediate its interaction with a variety of proteins including axin, adenomatous polyposis coli (APC), and T cell factor (TCF) family transcription factors. The C-terminal domain (CTD) of beta-catenin is implicated in transcriptional functions, while the N-terminal domain (NTD) contains several posttranslational modification (ubiquitination, acetylation, and phosphorylation) sites, including the Ser33/Ser37/Thr41 residues, important for beta-catenin stability regulation. In the absence of Wnt pathway activation, beta-catenin is retained in the cytoplasm in a “destruction complex” with Axin, APC, and GSK3beta, where it is constitutively destined for proteasomal degradation as a result of constant phosphorylation by GSK3beta. Activation of the Wnt pathway results in stabilization and nuclear translocation of beta-catenin, resulting in beta-catenin target genes transcription. In addition to GSK3beta, PKCalpha is also reported to phosphorylate the Ser33/37/Thr41 residues of beta-catenin, and beta-catenin Ser45 is a known phosphorylation site for CK1alpha. In addition, HDAC6 and p300/CBP-associated factor (PCAF) regulate beta-catenin Lys49 acetylation.

Specificity

Recognizes active form of β-catenin, dephosphorylated on Ser37 or Thr41.
The antibody eptiope is 100% conserved in zebrafish.

Immunogen

Epitope: The epitope corresponds to amino acid residues 36-44 (HSGATTTAP) of human β-catenin.
Fusion protein corresponding to residues 1-100 of human β-catenin with a c-terminal histidine tag.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Active-β-Catenin in A431 cells.
Flow Cytometry Analysis: 0.1 µg of this antibody from a representative lot detected Active-β-Catenin in 1X10E6 A431 and HeLa cells.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Active-β-Catenin in human colon and human colorectal adenocarcinoma tissue.
The unconjugated version (Cat. No. 05-665) has been shown to work in WB, FC, ICC, IHC, IH(P).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Active-β-Catenin Antibody, clone 8E7, Alexa Fluor 488 is validated for use in Immunocytochemistry, Flow Cytometry, Immunohistochemistry for the detection of Active-β-Catenin.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Active-β-Catenin in HeLa cells.

Target description

~85 kDa observed

Physical form

Protein G Purified
Purified mouse conjugate antibody in PBS with 15 mg/ml BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service