Skip to Content
Merck
All Photos(1)

Key Documents

05-23-1200

Sigma-Aldrich

Neurotensin

Potent hypothalamic peptide neurotransmitter.

Synonym(s):

Neurotensin, NT, NTS, Pyr-LYENKPRRPYIL

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€230.00

€230.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C78H121N21O20
CAS Number:
Molecular Weight:
1672.92
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€230.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N3(C(CCC3)C(=O)NC(CCCN=C(N)N)C(=O)NC(CCCN=C(N)N)C(=O)N4C(CCC4)C(=O)NC(Cc5ccc(cc5)O)C(=O)NC(C(CC)C)C(=O)NC(CC(C)C)C(=O)O)C(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C2NC(=O)CC2)CC(C)C)Cc1ccc(cc1)O)CCC(=O)O)CC(=O)N)CCCCN

InChI

1S/C78H121N21O20/c1-7-43(6)63(73(115)96-57(76(118)119)37-42(4)5)97-70(112)55(39-45-21-25-47(101)26-22-45)95-72(114)59-18-13-35-99(59)75(117)52(16-11-33-86-78(83)84)90-64(106)48(15-10-32-85-77(81)82)89-71(113)58-17-12-34-98(58)74(116)51(14-8-9-31-79)91-69(111)56(40-60(80)102)94-66(108)50(28-30-62(104)105)88-68(110)54(38-44-19-23-46(100)24-20-44)93-67(109)53(36-41(2)3)92-65(107)49-27-29-61(103)87-49/h19-26,41-43,48-59,63,100-101H,7-18,27-40,79H2,1-6H3,(H2,80,102)(H,87,103)(H,88,110)(H,89,113)(H,90,106)(H,91,111)(H,92,107)(H,93,109)(H,94,108)(H,95,114)(H,96,115)(H,97,112)(H,104,105)(H,118,119)(H4,81,82,85)(H4,83,84,86)

InChI key

PCJGZPGTCUMMOT-UHFFFAOYSA-N

General description

Neurotensin (NT), a 13-amino acid peptide neurotransmitter encoded by the NTS gene. NT is located in the gastrointestinal tract and heterogeneously distributed in the mammalian central nervous system, and has many neurotransmitter-like characteristics. It influences various central and peripheral functions by interacting with three neurotensin receptors. It is linked to the modulation of dopaminergic systems and in the induction of changes in behavior, muscle relaxation, nociception, and hypothermia[1], as well as appetite, learning, memory, and fat metabolism.[2] Moreover, it is a modulator of noradrenergic activity. It also acts as an endogenous anti-psychotic compound and lowers blood pressure, increases vascular permeability, and increases plasma levels of glucagon, glucose, growth hormone, and prolactin in rats. Additionally, it regulates the release of pituitary hormones from the brain and insulin, glucagon, and somatostatin from the gut.[3]

Biochem/physiol Actions

Cell permeable: no
Primary Target
Modulator of noradrenergic activity
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

Pyr-Leu-Tyr-Glu-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu-OH

Other Notes

Kinkead, B., et al. 1999. Biol. Psychiatr.46, 340.
Tyler, B.M., et al. 1998. Brain Res. 792, 246.
Tsuda, K., and Masuyuma, Y. 1993. Am. J. Hypertens. 6, 473.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V E Eysselein
Klinische Wochenschrift, 62(11), 523-530 (1984-06-01)
Neurotensin is a tridecapeptide originally isolated and characterized from bovine hypothalamus and later, in identical form, from bovine and human intestine. In the rat about 85% of immunoreactive neurotensin is found in the gut and about 10% in the brain.
Neurotensin Receptor Agents
Wendy Marsh
xPharm: The Comprehensive Pharmacology Reference, 1-3 (1007)
Napatsorn Saiyasit et al.
Neuropeptides, 72, 12-22 (2018-10-04)
Neurotensin is an endogenous tridecapeptide that can be found in both central and peripheral nervous systems. Under normal physiological conditions, neurotensin is involved in the regulation of pain, body temperature, physical activity, appetite as well as learning and memory. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service