Skip to Content
Merck
All Photos(2)

Documents

05-184-AF488

Sigma-Aldrich

Anti-Src Antibody, clone GD11, Alexa Fluor 488 conjugate

clone GD11, from mouse, ALEXA FLUOR 488

Synonym(s):

Tyrosine-protein kinase transforming protein Src, pp60v-src, p60-Src, v-Src, Proto-oncogene tyrosine-protein kinase Src, p60-Src, pp60c-src, Proto-oncogene c-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GD11, monoclonal

species reactivity

rat, human, mink, mouse, avian

technique(s)

immunocytochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

General description

Tyrosine-protein kinase transforming protein Src (EC 2.7.10.2; UniProt P00526; also known as p60-Src, pp60v-src, v-Src) is encoded by the V-SRC gene (Gene ID 1491925) of Rous sarcoma virus strain Prague C (RSV-PrC).
Rous sarcoma virus (RSV) V-SRC gene is the first retroviral oncogene discovered. V-SRC encodes the v-Src tyrosine kinase that transforms infected host cells into uncontrolled mitosis, providing abundant cells for fresh infection. The non-oncogenic chicken c-Src (cellular Src) gene structurally related to v-Src was discovered later in 1979 by J. Michael Bishop and Harold E. Varmus. It is believed that RSV originally took up the chicken proto-oncogene SRC (Gene ID 396442; UniProt P00523) and incorporated into its genome in the mutated concogene form. c-Src contains six functional regions, Src homology 4 (SH4) domain, unique region, SH3 domain, SH2 domain, catalytic domain, and short regulatory tail. In its inactive state, Src is phosphorylated at Tyr527. The pTyr527 interacts with the SH2 domain which helps the SH3 domain interact with the linker domain and consequently keeps the inactive unit tightly bound. Dephosphorylation of pTyr527 destabilizes the inactive conformation, opening up the SH3, SH2 and the kinase domains, leading to Tyr416 autophosphorylation and c-Src activation.

Specificity

Clone GD11 reacted with v-Src of Rous sarcoma virus (RSV) strains Prague subgroup A (PrA-RSV), Schmidt-Ruppin subgroup D (SRD-RSV), and Bratislava 77 subgroup C (B77 C-RSV). Clone GD11 detected enzymatically active c-Src from a variety of chicken, rodent, and human cells (Parsons, S.J., et al. (1984). J. Virol. 51(2):272-82).

Immunogen

Epitope: Amino acids 82-169.
Recombinant PrA-RSV p60src.

Application

Anti-Src Antibody, clone GD11, Alexa Fluor 488 conjugate is an antibody against Src for use in Immunocytochemistry.
Immunocytochemistry Analysis: An 1:100 dilution of this antibody from a representative lot detected Src in A431, HUVECs, and NIH/3T3 cells.
The unconjugated antibody (Cat. No. MAB1618) is shown to be suitable also for Western blotting, immunocytochemistry, immunofluorescence, and immunoprecipitation applications.
Research Category
Cell Structure
Research Sub Category
Oncoproteins

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: An 1:100 dilution of this antibody detected Src in HeLa cells.

Target description

~60 kDa observed.

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service