Skip to Content
Merck
All Photos(2)

Key Documents

05-1086

Sigma-Aldrich

Anti-phospho-IRS1 (Ser302 mouse/ Ser307 human) Antibody, clone 8.1.2

clone 8.1.2, from mouse

Synonym(s):

insulin receptor substrate 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€433.00

€433.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€433.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€433.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8.1.2, monoclonal

species reactivity

bovine, pig, canine, monkey, human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer302/pSer307)

Gene Information

bovine ... Irs1(538598)
dog ... Irs1(486148)
human ... IRS1(3667)
mouse ... Irs1(16367)
pig ... Irs1(100512686)
rat ... Irs1(25467)
rhesus monkey ... Irs1(707870)

General description

IRS1 (Insulin Receptor Substrate 1) transmits insulin signals via metabolic and mitogenic pathways. IRS1 is heavily phosphorylated on both serine and tyrosine residues. These phosphorylated tyrosines enable IRS to act as a docking protein that binds SH2 domains of such proteins as PI3 Kinase (phosphatidylinositol 3-kinase) and GRB2, resulting in activation. Over expression and phosphorylation of serine is associated with insulin resistance and breast cancer. Some of the more notable phosphorylation sites are Ser302 that is phosphorylated following insulin stimulation. Ser307, phosphorylated by JNK and IKK, is a key regulatory site that appears to disrupt the IRS1/IR interaction and inhibits insulin-mediated activation of the PI3 kinase and MAPK pathways, and Ser636/639 that is known to be phosphorylated by p70S6K downstream of mTOR and acts as a negative feedback loop.

Specificity

Predicted to cross-react with many other species based on 100% sequence homology with immunogen.
This antibody recognizes IRS1 phosphorylated at Ser302 (mouse) and Ser307 (human).

Immunogen

Epitope: Phosphorylated Ser302
Synthetic linear peptide corresponding to amino acids at and around phosphorylated Ser302 of mouse IRS1.

Application

Anti-phospho-IRS1 (Ser302 mouse/ Ser307 human) Antibody, clone 8.1.2 is an antibody against phospho-IRS1 (Ser302 mouse/ Ser307 human) for use in IH, IF, IP & WB.
Immunocytochemistry:
IR/IRS transfected CHO cells were stained with a 1:500 dilution of the antibody. A seconday anti-mouse Cy3 conjugate was used for visualizaion.

Immunoprecipition:
This antibody was used at a 1:1,000 to immunoprecipitate IRS1 from 5L/mL IR/IRS1 transfected CHO cells (performed by an independent lab).
Research Category
Metabolism

Signaling
Research Sub Category
Insulin/Energy Signaling

Glucose/Glycogen Metabolism

Metabolic Hormones & Receptors

Quality

Evaluated by western blot on IRS1 transfected CHO +/- Insulin lysate.

Western Blot Analysis:
1:1,000 dilution of this antibody was used to detect IRS1 phosphorylated on Ser302 in IRS/IR transfected CHO -/+ Calyculin A/ Okadaic Acid-treated cell lysate.

Target description

~185 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy J Hançer et al.
The Journal of biological chemistry, 289(18), 12467-12484 (2014-03-22)
IRS1 and IRS2 are key substrates of the insulin receptor tyrosine kinase. Mass spectrometry reveals more than 50 phosphorylated IRS1 serine and threonine residues (Ser(P)/Thr(P) residues) in IRS1 from insulin-stimulated cells or human tissues. We investigated a subset of IRS1
null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service