Skip to Content
Merck
All Photos(1)

Key Documents

02-103

Sigma-Aldrich

Bovine Pituitary Extract - 50 mg

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€462.00

€462.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€462.00

About This Item

UNSPSC Code:
41106200
eCl@ss:
32160801
NACRES:
NA.32

€462.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for cell culture

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

Application

For use as a cell culture media supplement.

Quality

routinely evaluated in a 5 day growth assay of fetal bovine heart endothelial cells

Physical form

Lyophilized from sterile water containing NaCl

Storage and Stability

Lyophilized: Stable for 2 years at 4°C from date of shipment. Rehydrated: Stable for 3 months at -20°C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service