Skip to Content
Merck
All Photos(1)

Documents

KEY158583362

Key Organics / BIONET

1-benzyl-3,5-dibromo-4-nitro-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
360.99
MDL number:

mp

132 - 133

Iupac Name

1-benzyl-3,5-dibromo-4-nitro-1H-pyrazole

Inchi Code

1S/C10H7Br2N3O2/c11-9-8(15(16)17)10(12)14(13-9)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

RNMYBBCIGXBZGA-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/NB-0821

Melting Point

132 - 133

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/NB-0821


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service