JWPH98890B08
J & W Pharmlab LLC
2-(2-Fluoro-phenyl)-1H-imidazo[4,5-c]pyridine methanesulfonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
309.32
Iupac Name
2-(2-fluorophenyl)-1H-imidazo[4,5-c]pyridine methanesulfonate
Inchi Code
1S/C12H8FN3.CH4O3S/c13-9-4-2-1-3-8(9)12-15-10-5-6-14-7-11(10)16-12;1-5(2,3)4/h1-7H,(H,15,16);1H3,(H,2,3,4)
InChI key
OVGLZVJGOQMWQA-UHFFFAOYSA-N
Purity
96%
Country of Origin
US
Physical Form
White Solid
Product Link
https://www.jwpharmlab.com/home/product/30R0061M
MSDS
https://www.jwpharmlab.com/home/product/30R0061M
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.