Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249DDA6

J & W Pharmlab LLC

3-Piperazin-1-yl-benzoic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
279.17
MDL number:

Iupac Name

3-(1-piperazinyl)benzoic acid dihydrochloride

Inchi Code

1S/C11H14N2O2.2ClH/c14-11(15)9-2-1-3-10(8-9)13-6-4-12-5-7-13;;/h1-3,8,12H,4-7H2,(H,14,15);2*1H

InChI key

YWKFNQMCJUDMHH-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Pink Solid

Product Link

https://www.jwpharmlab.com/home/product/65R0081S

MSDS

https://www.jwpharmlab.com/home/product/65R0081S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library