Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249C58A

J & W Pharmlab LLC

4-Dimethylamino-benzamidine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
236.14
MDL number:

Iupac Name

4-(dimethylamino)benzenecarboximidamide dihydrochloride

Inchi Code

1S/C9H13N3.2ClH/c1-12(2)8-5-3-7(4-6-8)9(10)11;;/h3-6H,1-2H3,(H3,10,11);2*1H

InChI key

QVWJLPLJSLCQMX-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Physical Form

Light Brown Solid

Product Link

https://www.jwpharmlab.com/home/product/12R0025S

MSDS

https://www.jwpharmlab.com/home/product/12R0025S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library