Skip to Content
Merck
All Photos(1)

Key Documents

ENAH95E73ED8

Enamine

(1-phenylethyl)[2-(pyrrolidin-1-yl)ethyl]amine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
291.26
MDL number:

Iupac Name

1-phenyl-N-(2-(pyrrolidin-1-yl)ethyl)ethan-1-amine dihydrochloride

Inchi Code

1S/C14H22N2.2ClH/c1-13(14-7-3-2-4-8-14)15-9-12-16-10-5-6-11-16;;/h2-4,7-8,13,15H,5-6,9-12H2,1H3;2*1H

InChI key

NYBUVRRUAVAFMH-UHFFFAOYSA-N

Salt Data

Cl

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-27159048

MSDS

https://www.enaminestore.com/catalog/EN300-27159048


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library