Skip to Content
Merck
All Photos(1)

Key Documents

ENA238710095

Enamine

4-(2-sulfanyl-1H-imidazol-1-yl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.25
MDL number:

mp

338-340

Iupac Name

4-(2-thioxo-2,3-dihydro-1H-imidazol-1-yl)benzoic acid

Inchi Code

1S/C10H8N2O2S/c13-9(14)7-1-3-8(4-2-7)12-6-5-11-10(12)15/h1-6H,(H,11,15)(H,13,14)

InChI key

ZNFFXFYIHZSXFP-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-42843

Melting Point

338-340

MSDS

https://www.enaminestore.com/catalog/EN300-42843


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library