Skip to Content
Merck
All Photos(1)

Key Documents

COM490021317

Combi-Blocks, Inc.

Perfluorohexanoyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
316.05
MDL number:

Iupac Name

2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoyl fluoride

Inchi Code

1S/C6F12O/c7-1(19)2(8,9)3(10,11)4(12,13)5(14,15)6(16,17)18

InChI key

XATLHBQMSOZWBO-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QF-5064

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QF-5064


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library