Skip to Content
Merck
All Photos(1)

Key Documents

CLE969755522

Clearsynth

6-Mercaptopurine-13C2 15N

1,9-Dihydro-6H-purine

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
155.16

Iupac Name

7H-purine-6-thiol-4,8-13C2-7-15N

Inchi Code

1S/C5H4N4S/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)/i1+1,4+1,6+1

InChI key

GLVAUDGFNGKCSF-JDKPDMQJSA-N

storage temp.

Normal Condition

Purity

99.87

Country of Origin

CA

Product Link

http://www.clearsynth.com/en/CSO02871.html

MSDS

http://www.clearsynth.com/en/CSO02871.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library