CIAH98ABF3A3
ChemScene LLC (Preferred Partner)
2-Chloro-4-(trifluoromethyl)thiazole-5-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
337.4+-52.0 C at 760 mmHg
mp
131-135 C
Iupac Name
2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Inchi Code
1S/C5HClF3NO2S/c6-4-10-2(5(7,8)9)1(13-4)3(11)12/h(H,11,12)
InChI key
OGZUWSFEZCBGPH-UHFFFAOYSA-N
storage temp.
4C
Purity
97%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/72850-61-4.html
Boiling point
337.4+-52.0 C at 760 mmHg
Melting Point
131-135 C
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0060012.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service