CIAH9884F700
ChemScene LLC (Preferred Partner)
1,4-Bis(2-methyl-1H-imidazol-1-yl)benzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
238.29
Iupac Name
1,4-bis(2-methyl-1H-imidazol-1-yl)benzene
Inchi Code
1S/C14H14N4/c1-11-15-7-9-17(11)13-3-5-14(6-4-13)18-10-8-16-12(18)2/h3-10H,1-2H3
InChI key
IIYKBRHJIZFDAJ-UHFFFAOYSA-N
storage temp.
4C
Purity
98%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/1347702-71-9.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0110398.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.