Skip to Content
Merck
All Photos(1)

Documents

U-003

Supelco

U-47700 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

U-47700 reference solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22Cl2N2O
Molecular Weight:
329.26
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

shipped in

wet ice

storage temp.

−20°C

General description

U-47700 is a highly potent opioid analgesic drug that has been confirmed in many opioid overdoses and deaths and was found in combination with fentanyl during the autopsy of the American guitarist Prince in 2016. U-47700 is regulated as a Schedule I compound in the United States. This Certified Spiking Solution® is applicable for use in urine drug testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.

This Cerilliant product is part of the Centers for Disease Control and Prevention (CDC) Opioid CRM Kit including more than 40 CRMs to be used as part of CDC′s laboratory response to the opioid epidemic. The kits are provided free of charge to drug testing laboratories located in the United States. A requesting lab must have current DEA registration and must comply with all respective state and local regulations. Visit www.cerilliant.com for instructions on requesting a free Opioid CRM Kit. Please note, CDC will review and approve all order requests prior to shipment.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service