Skip to Content
Merck
All Photos(1)

Key Documents

T-118

Supelco

Trifluoperazine N-ß-D-glucuronide solution

100 μg/mL in methanol: water (1:1), ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32F3N3O6S
CAS Number:
Molecular Weight:
583.62
UNSPSC Code:
12352200
NACRES:
NA.24
Pricing and availability is not currently available. Contact Technical Service

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

InChI

1S/C27H32F3N3O6S/c1-33(25-23(36)21(34)22(35)24(39-25)26(37)38)13-11-31(12-14-33)9-4-10-32-17-5-2-3-6-19(17)40-20-8-7-16(15-18(20)32)27(28,29)30/h2-3,5-8,15,21-25,34-36H,4,9-14H2,1H3/t21-,22-,23+,24-,25+/m0/s1

InChI key

AFYPHWNGUDUHIW-ARXROMJUSA-N

General description

Trifluoperazine N-β-D-glucuronide is a urinary and blood metabolite of trifluoperazine, a typical antipsychotic used for treating schizophrenia. Trifluoperazine is also indicated for use in patients with agitation, severe anxiety, or behavioral problems. This Snap-N-Spike®; reference material is suitable for use as a hydrolysis control in mass spectrometry-based testing applications of trifluoperazine in clinical toxicology, forensic analysis, therapeutic drug monitoring, and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service