Skip to Content
Merck
All Photos(1)

Documents

M-209

Supelco

Milnacipran-D10 hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13D10ClN2O
CAS Number:
Molecular Weight:
292.87
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(N(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H])[C@]1(C2=CC=CC=C2)[C@@H](C1)CN.Cl

General description

This Snap-N-Shoot® stable-labeled internal standard is suitable for quantitation of milnacipran levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis. Milnacipran, sold under the brand names Savella, Ixel, Dalcipran, or Toledomin, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the treatment of depression. In 2009, the US Food and Drug Administration (FDA) approved milnacipran for management of the chronic pain disorder fibromyalgia, making it only the third drug approved in the US for this indication besides duloxetine and pregabalin.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service