Skip to Content
Merck
All Photos(1)

Key Documents

I-011

Supelco

Iloperidone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27FN2O4
CAS Number:
Molecular Weight:
426.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CC(C1=CC=C(OCCCN2CCC(C3=NOC4=C3C=CC(F)=C4)CC2)C(OC)=C1)=O

InChI

1S/C24H27FN2O4/c1-16(28)18-4-7-21(23(14-18)29-2)30-13-3-10-27-11-8-17(9-12-27)24-20-6-5-19(25)15-22(20)31-26-24/h4-7,14-15,17H,3,8-13H2,1-2H3

InChI key

XMXHEBAFVSFQEX-UHFFFAOYSA-N

Gene Information

General description

Iloperidone is an atypical antipsychotic administered for the treatment of schizophrenia. The drug is marketed under the trade names Fanapt and Fanapta. This Certified Spiking Solution® is suitable for use in LC/MS or GC/MS applications including clinical toxicology, forensic testing, or pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Weiden
Clinical schizophrenia & related psychoses, 6(1), 34-44 (2012-03-29)
Iloperidone is a second-generation "atypical" antipsychotic whose primary mechanism of action is within the subclass of combined D2/5HT2A antagonism. Iloperidone was approved by the FDA in May 2009 for the treatment of schizophrenia. This review is a comprehensive synthesis of
Heidi N Boyda et al.
PloS one, 8(1), e53459-e53459 (2013-01-18)
The second generation antipsychotic (SGA) drugs are widely used in psychiatry due to their clinical efficacy and low incidence of neurological side-effects. However, many drugs in this class cause deleterious metabolic side-effects. Animal models accurately predict metabolic side-effects for SGAs
Bonnie A Fijal et al.
The Journal of clinical psychiatry, 73(3), 367-371 (2011-08-05)
We examined 6 single nucleotide polymorphisms (SNPs) previously reported to be associated with response to iloperidone therapy for association with response to risperidone therapy. Patients with schizophrenia (DSM-IV) were assessed during 2006 and 2007 for response/nonresponse (defined as ≥ 20%/<20%
Radha Charan Dash et al.
Molecular diversity, 16(2), 367-375 (2011-12-14)
The dopamine D(2) receptor is involved in the etiology of a number of disorders, such as Parkinson's disease, Huntington's Chorea, tardive dyskinesia and schizophrenia. Antagonism of D(2) receptors is implicated in the treatment of various psychiatric disorders. In order to
Mengmeng Jia et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 928, 52-57 (2013-04-27)
A selective, sensitive and accurate high-performance liquid chromatographic- tandem mass spectrometry (HPLC-MS/MS) method for simultaneous determination of iloperidone and its two active metabolites, P88 and P95, in human plasma has been first developed and validated. The analytes and internal standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service