880136P
Avanti
DSPE-PEG(2000)-square
Avanti Research™ - A Croda Brand (880136P), powder
Synonym(s):
1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[square(polyethylene glycol)-2000] (sodium salt)
About This Item
Recommended Products
Assay
99% (TLC)
form
powder
packaging
pkg of 1 × 10 mg (880136P-10mg)
pkg of 1 × 25 mg (880136P-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand (880136P)
shipped in
dry ice
storage temp.
−20°C
General description
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service