Skip to Content
Merck
All Photos(1)

Key Documents

852478P

Avanti

C16(Plasm)-18:1 PC

Avanti Research - A Croda Brand

Synonym(s):

1-(1Z-haxadecenyl)-2-oleoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€331.00

€331.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€331.00

About This Item

Empirical Formula (Hill Notation):
C42H82NO7P
CAS Number:
Molecular Weight:
744.08
UNSPSC Code:
12352211
NACRES:
NA.21

€331.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (852478P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

General description

C16(Plasm)-18:1 PC, also known as 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, is a phosphatidylcholine (PC), which is a type of phospholipid. Phospholipids are major components of cell membranes and play vital roles in membrane structure and function.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service