Skip to Content
Merck
All Photos(2)

Key Documents

850700C

Avanti

10:0 PE

1,2-didecanoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

PE(10:0/10:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€166.00

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€166.00

About This Item

Empirical Formula (Hill Notation):
C25H50NO8P
CAS Number:
Molecular Weight:
523.64
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850700C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850700C

concentration

10 mg/mL (850700C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCC)=O)COC(CCCCCCCCC)=O

InChI

1S/C25H50NO8P/c1-3-5-7-9-11-13-15-17-24(27)31-21-23(22-33-35(29,30)32-20-19-26)34-25(28)18-16-14-12-10-8-6-4-2/h23H,3-22,26H2,1-2H3,(H,29,30)/t23-/m1/s1

InChI key

KKOSJVWUOHEQKA-HSZRJFAPSA-N

General description

1,2-didecanoyl-sn-glycero-3-phosphoethanolamine (10:0 PE) is a soluble short-chain phospholipid.[1]

Application

1,2-didecanoyl-sn-glycero-3-phosphoethanolamine (10:0 PE) may be used as an internal standard for the quantitative analysis of phospholipids by electrospray ionization-tandem mass spectrometry (ESI-MS/MS).[2] It may also be used to titrate the lantibiotic cinnamycin (Ro 09-0198) in isothermal titration calorimetry.[3]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) controls membrane fluidity in eukaryotic cells.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850700C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Cortese et al.
Biochemistry, 26(17), 5283-5293 (1987-08-25)
D-beta-Hydroxybutyrate dehydrogenase (BDH) is a lecithin-requiring mitochondrial enzyme that catalyzes the interconversion of beta-hydroxybutyrate and acetoacetate. The purified soluble enzyme devoid of lipid (i.e., the apodehydrogenase) can be reactivated with soluble lecithin or by insertion into phospholipid vesicles containing lecithin.
Gia Machaidze et al.
Biochemistry, 42(43), 12570-12576 (2003-10-29)
Cinnamycin (Ro 09-0198) is a tetracyclic peptide antibiotic that binds specifically to phosphatidylethanolamine (PE). Formation of a complex with phosphatidylethanolamine follows a 1:1 stoichiometry. Using high-sensitivity isothermal titration calorimetry (ITC), we have measured the thermodynamic parameters of complex formation for
Shohei Kotani et al.
Fungal genetics and biology : FG & B, 96, 1-11 (2016-10-22)
In mammals, cytosolic phospholipases A2 (cPLA2s) play important physiological roles by releasing arachidonic acid, a precursor for bioactive lipid mediators, from the biological membranes. In contrast, fungal cPLA2-like proteins are much less characterized and their roles have remained elusive. AoPlaA
Rosie Dawaliby et al.
The Journal of biological chemistry, 291(7), 3658-3667 (2015-12-15)
Adequate membrane fluidity is required for a variety of key cellular processes and in particular for proper function of membrane proteins. In most eukaryotic cells, membrane fluidity is known to be regulated by fatty acid desaturation and cholesterol, although some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service