Skip to Content
Merck
All Photos(1)

Key Documents

700168P

Avanti

16:0 stigmasteryl glucose

6-O-palmitoyl-stigmasteryl-β-D-glucose, powder

Synonym(s):

3-O-[(6′-O-palmitoyl)-β-D-glucopyranosyl] stigmasterol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€162.00

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C51H88O7
CAS Number:
Molecular Weight:
813.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700168P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700168P

shipped in

dry ice

storage temp.

−20°C

General description

Stigmasteryl glucoside (SG) comprises the glucose attached to the C3-hydroxyl of the sterol backbone. The acylstigmasteryl glucoside has a fatty acid at the C6 position of glucose.[1] Palmitic acid is the acyl component in 16:0 stigmasteryl glucose and is a predominant acyl group in Arabidopsis seeds.[1]

Biochem/physiol Actions

The levels of the 16:0 (palmitic) and other acyl steryl glycosides (ASG) are depleted when there is a mutation in the DP-glucose:sterol glucosyltransferase (UGT) enzyme.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700168P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathrin Schrick et al.
Lipids, 47(2), 185-193 (2011-08-11)
Establishment of sensitive methods for the detection of cellular sterols and their derivatives is a critical step in developing comprehensive lipidomics technology. We demonstrate that electrospray ionization tandem (triple quadrupole) mass spectrometry (ESI-MS/MS) is an efficient method for monitoring steryl
Daniel F Stucky et al.
Journal of experimental botany, 66(1), 189-201 (2014-10-16)
Steryl glucosides (SG) are abundant steroid conjugates in plant membranes. Beyond structural roles in lipid bilayers, functions in sugar transport, storage, and/or signalling are predicted. UDP-glucose:sterol glucosyltransferase 80A2 (UGT80A2) and UGT80B1, which share similarity to fungal counterparts, are implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service