Skip to Content
Merck
All Photos(2)

Key Documents

700043P

Avanti

7α-hydroxycholesterol-d7

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3b,7a-diol(d7); 110971

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€791.00

€791.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€791.00

About This Item

Empirical Formula (Hill Notation):
C27H39O2D7
CAS Number:
Molecular Weight:
409.70
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

€791.00


Please contact Customer Service for Availability

Request a Bulk Order

description

cholest-5-en-3β,7α-diol-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700043P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Application

7α-hydroxycholesterol-d7 has been used as an internal standard for cholesterol and oxysterol measurement by gas chromatography-mass spectrometry (GC/MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700043P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aerobic exercise training selectively changes oxysterol levels and metabolism reducing cholesterol accumulation in the aorta of Dyslipidemic mice
Ferreira GS, et al.
Frontiers in Physiology, 44(9), 644-644 (2017)
Alzbeta Kloudova-Spalenkova et al.
The Journal of steroid biochemistry and molecular biology, 197, 105566-105566 (2019-12-25)
Oxygenated metabolites of cholesterol (oxysterols) have been previously demonstrated to contribute to progression of various cancers and to modulate resistance to breast cancer endocrine therapy in vitro. We measured prognostic roles of circulating levels of seven major oxysterols in the
Louise Ménégaut et al.
Cell reports, 31(7), 107665-107665 (2020-05-21)
Low-grade inflammation is constitutive of atherosclerosis, and anti-inflammatory therapy inhibiting interleukin-1β (IL-1β) reduces the rate of cardiovascular events. While cholesterol accumulation in atheroma plaque and macrophages is a major driver of the inflammatory process, the role of the LXR cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service