Skip to Content
Merck
All Photos(1)

Key Documents

ATEH03AB15A3

AstaTech, Inc.

PYRIDIN-2-YLMETHANESULFONIC ACID HCL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
209.65
MDL number:
Pricing and availability is not currently available.

Iupac Name

pyridin-2-ylmethanesulfonic acid hydrochloride

Inchi Code

1S/C6H7NO3S.ClH/c8-11(9,10)5-6-3-1-2-4-7-6;/h1-4H,5H2,(H,8,9,10);1H

InChI key

ZVTAJMFYZISBNI-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=115944

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=95731


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library