ATE370310780
AstaTech, Inc.
1-METHYLCYCLOHEXANECARBALDEHYDE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-methylcyclohexanecarbaldehyde
Inchi Code
1S/C8H14O/c1-8(7-9)5-3-2-4-6-8/h7H,2-6H2,1H3
InChI key
FXZFFVCJWZTTMX-UHFFFAOYSA-N
storage temp.
-20 C
Purity
85%
Country of Origin
CN
Shipping Temperature
Cold box
Physical Form
Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=10910
Flash Point
51 C/123.8 F
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=BL001447
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service