APO456796935
Apollo Scientific Ltd
2-Fluoro-5-(trifluoromethyl)benzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
115-117
Iupac Name
2-fluoro-5-(trifluoromethyl)benzamide
Inchi Code
1S/C8H5F4NO/c9-6-2-1-4(8(10,11)12)3-5(6)7(13)14/h1-3H,(H2,13,14)
InChI key
DDLPGTOHZZKQLP-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzamide
Synonym
3-Carbamoyl-4-fluorobenzotrifluoride, 3-(Aminocarbonyl)-4-fluorobenzotrifluoride
Boiling point
115-117
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzamide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.