Iupac Name
methyl 3-bromopropiolate
Inchi Code
1S/C4H3BrO2/c1-7-4(6)2-3-5/h1H3
InChI key
PITMUHRRCBFULF-UHFFFAOYSA-N
storage temp.
Sealed in dry,Store in freezer, under -20C
Purity
95%
Country of Origin
CN
Shipping Temperature
Shipped in cold pack
Physical Form
Colorless to White to yellow to brown Liquid or Solid
Product Link
https://www.ambeed.com/products/23680-40-2.html
Signal Word
Danger
Hazard Statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.