Z742344
2MAG steriMIXdrive, Steam, pressure and vacuum resistant stirrer for viscous material
use with steriMIXcontrol unit
Synonym(s):
induction plate, induction stirrer, laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
Select a Size
€1,880.00
Select a Size
About This Item
€1,880.00
Recommended Products
AC/DC input
24 V
material
stainless steel (PPSU, hermetically closed)
stainless steel plate
feature
positions 1
packaging
pack of 1 ea
manufacturer/tradename
2mag 40700
parameter
-10-150 °C temp. limit (at 100% humidity)
100-1200 rpm speed
W × D × H
140 mm × 140 mm × 32 mm
plate size × W × L
× 140 mm × 140 mm
stirrer volume
1-40 L , per stirring position
weight
9.0 kg
General description
stirring volume, also suitable for highly viscous media.
Very strong magnetic coupling by SmCo-magnets, therefore
predestined for magnetic power transmission over wide distances or for thick vessel bottoms. Maintenance-free 2mag-motor-drive with large speed range from 100 up to 1,200 rpm.
Fully encapsulated, pressure- and vacuum resistant stainless steel housing, IP68, temperature resistant up to +150 C, extremely robust construction and performance, easy to clean and firm.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service