material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
3 kDa
packaging
pack of 24 ea
manufacturer/tradename
Millipore UFC200324
tube size
~2 mL
working volume
2 mL
pore size
3 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)