Z696196
Whatman® qualitative filter paper for technical use, Grade 2294
circles, diam. 110 mm, pack of 100
Synonym(s):
Whatman filter, Whatman paper, paper filter
About This Item
Recommended Products
material
cellulose filters (super-refined)
circles
packaging
pack of 100
manufacturer/tradename
Whatman 10342810
Whatman Article No. 28414578 (US reference)
parameter
27.5 sec/100 mL speed (Herzberg)
diam.
110 mm
thickness
1500 μm
pore size
8-15 μm (Particle retention)
basis weight
556 g/m2
General description
Grade 2294: 8-15 μm
A very thick filter card with high wet strength. Offers very high flow rate and retains medium to coarse particles.
Other Notes
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service