Key Documents
Z680559
Caps for Duran® laboratory bottles
neck thread for GL 45, blue-yellow polypropylene tamper-evident (with lip seal)
Synonym(s):
caps bottles, glass bottles caps
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Product Z680559 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
DURAN®
Schott
fitting
neck thread for GL 45
General description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)