Skip to Content
Merck
All Photos(1)

Documents

Z676187

Plum pH neutralizing eyewash refill

pkg of 200 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

packaging

pkg of 200 mL

manufacturer/tradename

Plum

General description

A small handy bottle containing 4.9% sterile phosphate buffer solution, which neutralizes the eye′s fluid after accidents involving acids or alkalis. The bottle is easy to take with you or can be used to refill the neutralizing eyewash station. The bottle is provided with an ergonomic eyecup, dust cap and clear instructions.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service