Documents
Z660612
Plas-Labs temperature and humidity control box with drying train
AC/DC input 240 V AC (EU plug)
Synonym(s):
isolation chambers, pcr chambers
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
manufacturer/tradename
Plas-Labs 890-THC/DT/EXP/EU
inside H × W × D
26 in. × 41 in. × 28 in.
outside H × W × D
38 in. × 55 in. × 35 in.
General description
Price includes shipping
The Plas-Labs temperature and humidity control glove box is designed to maintain accurate control of the isolated atmosphere. The drying train can be used when in-house inert gas is not available. The molecular sieve should be regenerated periodically for optimal drying performance.
The easy to use touch screen offers password protection, on screen readings of chamber temperature and humidity, full color screen, and room temperature and humidity readings.
  • Humidity (Rh) transmitter with 8ft. sensor cable
  • Two inlet/outlet gas valves on main chamber and transfer chamber
  • Serial port connection for data logging
  • Thermoelectric assembly (Peltier device)
  • Nebulizer
  • Two drying train canisters (filled with Molecular Sieve) and circulation pump
  • CE compliant