Key Documents
Z654892
Best® Nitri-Care® gloves
size 7‑8 M
About This Item
UNSPSC Code:
46181504
material
nitrile
size
7‑8 M
sterility
non-sterile
feature
CE compliant
disposable
latex free
powder-free
packaging
package of 100 ea
manufacturer/tradename
Best Manufacturing 3005PFM
L × thickness
9.5 in. × 4 mil
color
blue
General description
  • Tested according to EN 455
  • Ambidextrous
  • The "low-modulus" feature: the glove becomes a second skin
  • Nitri-Care provides three times the puncture resistance of natural rubber and vinyl gloves of comparable thickness (puncture resistance is > 0.45 Kg)
  • Best protection against latex allergies, viruses and bacteria
Nitri-Care is the economical medical examination version of the N-DEX family. Made in ISO 9002 quality-certified facility, Nitri-Care is inspected to a quality level of 1.5AQL. Nitri-Care does not contain any proteins of natural rubber and features the proprietary non-allergenic polymeric coating that aids in wet-hand donning.
  • Tested according to EN 455
  • Ambidextrous
  • The "low-modulus" feature: the glove becomes a second skin
  • Nitri-Care provides three times the puncture resistance of natural rubber and vinyl gloves of comparable thickness (puncture resistance is > 0.45 Kg)
  • Best protection against latex allergies, viruses and bacteria
Legal Information
Best is a registered trademark of Showa Glove Co.
Nitri-Care is a registered trademark of Best Manufacturing Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)