Documents
Z303216
Quickfit® Liebig condenser
L 400 mm, joint: ST/NS 24/29
Synonym(s):
C1/23/SC
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/29
manufacturer/tradename
Quickfit C1/23/SC
availability
not available in USA and Canada
not available in USA and Canada
L
400 mm
surface area
200 cm2
General description
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Legal Information
Quickfit is a registered trademark of SciLabware Limited