Documents
Z300969
Quickfit® Reduction Adapters
top female joint: ST/NS 24/29, bottom male joint: ST/NS 29/32
Synonym(s):
DA34
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS top24/29
male joint: ST/NS bottom29/32
manufacturer/tradename
Quickfit DA 34
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited