Skip to Content
Merck
All Photos(1)

Key Documents

Z264881

Sigma-Aldrich

Hydrion® pH Jumbo dispenser

pH 1-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034

feature

non-bleeding: no
refill: no

manufacturer/tradename

Hydrion

graduations and accuracy measuring range

accuracy: 1 pH unit

pH measuring range

1-12

W × L

1/2 in. × 50 ft

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

Jumbo size paper is easier to read with large size color charts. Yields a sharp color change for each pH unit.

Legal Information

Hydrion is a registered trademark of Micro Essential Laboratory, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service