Documents
Z107115
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 75/50
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
stainless steel
Joint
joint: SJ75/50
General description
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.